Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Propylene Oxide, EM Grade 250 ML
SDP

Catalog No. 5028186
Encompass_Preferred

(1,2-Epoxypropane, Methyloxirane)
CAS #75-56-9 CH₃CHCH₂O F.W. 58.07
Melting point: -112°C
Boiling point: 34°C

A solvent agent for epoxy resins. Used widely for infiltration and gradual replacement of the dehydration agent.
Grade: EM Grade

Catalog No. 50-281-86 Supplier Electron Microscopy Sciences Supplier No. 20414
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1,2-Epoxypropane, Methyloxirane)
CAS #75-56-9 CH₃CHCH₂O F.W. 58.07
Melting point: -112°C
Boiling point: 34°C

A solvent agent for epoxy resins. Used widely for infiltration and gradual replacement of the dehydration agent.
Grade: EM Grade

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.