Learn More
Electron Microscopy Sciences Propylene Oxide, EM Grade 4x450 ML Metal Cans

Supplier: Electron Microscopy Sciences 20411
(1,2-Epoxypropane, Methyloxirane)
CAS #75-56-9 CHCHCHO F.W. 58.07
Melting point: -112C
Boiling point: 34C
A solvent agent for epoxy resins. Used widely for infiltration and gradual replacement of the dehydration agent.
Grade: EM Grade
4x450 ml Metal Cans
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.