Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene sulfide, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1072-43-1
C3H6S
74.141
MFCD00005160
MBNVSWHUJDDZRH-UHFFFAOYSA-N
propylene sulfide,thiirane, methyl,thiirane, 2-methyl,propene sulfide,1,2-epithiopropane,propylene sulphide,propylene episulfide,2-methylthiacyclopropane,propane, 1,2-epithio,methylthiirane
14072
2-methylthiirane
CC1CS1
1072-43-1
C3H6S
74.141
MFCD00005160
MBNVSWHUJDDZRH-UHFFFAOYSA-N
propylene sulfide,thiirane, methyl,thiirane, 2-methyl,propene sulfide,1,2-epithiopropane,propylene sulphide,propylene episulfide,2-methylthiacyclopropane,propane, 1,2-epithio,methylthiirane
14072
2-methylthiirane
CC1CS1
Density 0.94g/mL
Refractive Index 1.4740 to 1.4780 (20°C, 589nm)
Packaging Glass bottle
Quantity 100g
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 96%
UN Number 1992
Beilstein 17, II, 15
Formula Weight 74.14
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material Propylene sulfide
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

Danger