Learn More
U.S. Pharmacopeia Propylparaben, 94-13-3, MFCD00002354, 200mg
Catalog No. NC9197583
Encompass
Molecular formula C10H12O3, Molecular weight 180.2, Melting Point 203 - 210.2 °F (95 - 99 °C), Flash point 356.00 °F (180.00 °C) (method not specified), Synonyms: Propyl p-hydroxybenzoate
Catalog No. NC9197583 Supplier U.S. Pharmacopeia Supplier No. 1577008
Molecular formula C10H12O3, Molecular weight 180.2, Melting Point 203 - 210.2 °F (95 - 99 °C), Flash point 356.00 °F (180.00 °C) (method not specified), Synonyms: Propyl p-hydroxybenzoate