Learn More
Cayman Chemical Propylparaben, Purity: ≥98%, CAS: 94-13-3, 5 g
SDP
Catalog No. 502918218
Encompass_Preferred
A paraben with diverse biological activities; active against several strains of P. stutzeri (MICs = 250-300 µg/ml); binds to estrogen receptors in a concentration-dependent manner; decreases sperm counts and serum testosterone levels in rats at 1% in the diet; has been found in influent and effluent wastewater
Catalog No. 50-291-8218 Supplier Cayman Chemical Supplier No. 374175 G
A paraben with diverse biological activities; active against several strains of P. stutzeri (MICs = 250-300 µg/ml); binds to estrogen receptors in a concentration-dependent manner; decreases sperm counts and serum testosterone levels in rats at 1% in the diet; has been found in influent and effluent wastewater