Learn More
Medchemexpress LLC Propylparaben-d4 | 1219802-67-1 | 184.23 | 1 MG
SDP
Catalog No. 5000415940
Encompass_Preferred
Propylparaben-d4 is the deuterium-labeled version of Propylparaben. Propylparaben, also known as Propyl parahydroxybenzoate, is a naturally occurring antimicrobial preservative found in plants and bacteria. It is widely used in various products, including cosmetics, pharmaceuticals, and foods.
  • Deuterium labeled: This compound is enriched with deuterium, making it suitable for specific research applications.
  • Antimicrobial preservative: Its unlabeled counterpart acts as an antimicrobial preservative.
  • Research applications: Can be used as a tracer and as an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS.
  • Impact on biological systems: Studies indicate that Propylparaben can disrupt antral follicle growth and steroidogenic function by altering cell-cycle, apoptosis, and steroidogenesis pathways. It has also been shown to decrease sperm count and motility in rats.
Catalog No. 50-004-15940 Supplier Medchemexpress LLC Supplier No. HYN2026S11MG
Propylparaben-d4 is the deuterium-labeled version of Propylparaben. Propylparaben, also known as Propyl parahydroxybenzoate, is a naturally occurring antimicrobial preservative found in plants and bacteria. It is widely used in various products, including cosmetics, pharmaceuticals, and foods.
  • Deuterium labeled: This compound is enriched with deuterium, making it suitable for specific research applications.
  • Antimicrobial preservative: Its unlabeled counterpart acts as an antimicrobial preservative.
  • Research applications: Can be used as a tracer and as an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS.
  • Impact on biological systems: Studies indicate that Propylparaben can disrupt antral follicle growth and steroidogenic function by altering cell-cycle, apoptosis, and steroidogenesis pathways. It has also been shown to decrease sperm count and motility in rats.