Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Propylphosphonic anhydride solution

Catalog No. 81801100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-801-100ML 100 mL
81-801-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 81-801-100ML Supplier Sigma Aldrich Supplier No. 81801100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Propylphosphonic anhydride is a highly reactive cyclic anhydride that is commonly used as a coupling agent and water scavenger.Coupling reagent for peptide synthesis; Preparation of other amides; Synthesis of esters (depsipeptides)T3P is a registered trademark of Archimica GmbH

Specifications

CAS 68957-94-8
Molecular Formula C9H21O6P3
Linear Formula C9H21O6P3
Quantity 100 mL
MDL Number MFCD00006583
Synonym 1-Propanephosphonic anhydride solution; 2,4,6-Tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide solution; PPACA; T3P™
Molecular Weight (g/mol) 318.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.