Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Propylphosphonic anhydride solution

Catalog No. 43130350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
100 mL
500 mL
2.5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-130-350ML 50 mL
43-130-325L 2.5 L
43-130-3100ML 100 mL
43-130-310ML 10 mL
43-130-3500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 43-130-350ML Supplier Sigma Aldrich Supplier No. 43130350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Propylphosphonic anhydride (t3p) is a reactive n-propyl phosphonic acid cyclic anhydride. it is a mild and low toxic coupling agent used in peptide synthesis. t3p also acts as a promoter and water scavenger in the Friedländer annulation reaction. it participates in the conversion of carboxylic acids and amides into nitriles, formation of weinreb amides, ester synthesis, dehydrations, oxidation of alcohols, isonitrile synthesis, synthesis of alkenes from alcohols and c-c coupling reactions. t3p delivers outstanding advantages over traditional reagents, such as broad functional group tolerance, low epimerization, and water-soluble by-products and hence gives high purity and yield of the product.propylphosphonic anhydride may be used in the following studies:
  • As coupling agent for the synthesis of bispyridine-based ligands, which are used as bridging linkers in multinuclear platinum anticancer drugs
  • Microwave-assissted fischer indolization of arylhydrazines
  • As acid activating agent for the direct synthesis of acid azides from carboxylic acids
  • One-pot synthesis of coumarins
  • Microwave-mediated synthesis of carbocyclic and heterocyclic fused quinolones
  • One-pot synthesis of 1,2,4-oxadiazoles, 1,3,4-oxadiazoles, and 1,3,4-thiadiazoles from carboxylic acids
  • Activation of the carboxyl group for hydroxyamidation and peptide coupling and in the one-pot conversion of carboxylic acids into hydroxamic acids.t3p is a registered trademark of archimica gmbh

Specifications

CAS 68957-94-8
Density 1.069 g/mL (at 25°C)
Boiling Point 65°C
Molecular Formula C9H21O6P3
Linear Formula C9H21O6P3
Refractive Index n20/D 1.418
MDL Number MFCD00006583
Quantity 50 mL
Synonym 1-Propanephosphonic anhydride solution; 2,4,6-Tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide solution; PPACA; T3P™
Molecular Weight (g/mol) 318.18
Biological Activity Central Nervous System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.