Learn More
Cayman Chemical Propylpyrazole Triol, Purity: ≥98%, CAS: 263717-53-9, 25 mg
SDP
Catalog No. 502890001
Encompass_Preferred
An ERα selective agonist with a 410-fold relative binding affinity for ERα versus ERβ
Catalog No. 50-289-0001 Supplier Cayman Chemical Supplier No. 1000884125 MG
An ERα selective agonist with a 410-fold relative binding affinity for ERα versus ERβ