Learn More
Cayman Chemical PropylthIouracIl-d5 2.5mg
SDP
Catalog No. 502990241
Encompass_Preferred
An internal standard for the quantification of propylthiouracil by GC- or LC-MS
Catalog No. 50-299-0241 Supplier Cayman Chemical Supplier No. 307432.5 MG
An internal standard for the quantification of propylthiouracil by GC- or LC-MS