Learn More
Selleck Chemical LLC Propylthiouracil 10mg 51-52-5 NSC 6498, NSC 70461
SDP
Catalog No. 506950
Encompass_Preferred
Propylthiouracil (NSC 6498, NSC 70461) is a thyroperoxidase and 5'-deiodinase inhibitor, used to treat hyperthyroidism. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-695-0 Supplier Selleck Chemical LLC Supplier No. S198810MG
Propylthiouracil (NSC 6498, NSC 70461) is a thyroperoxidase and 5'-deiodinase inhibitor, used to treat hyperthyroidism. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.