Learn More
Medchemexpress LLC Propynyl-PEG1-Ac 50mg
SDP
Catalog No. 5000205794
Encompass_Preferred
Propynyl-PEG1-Ac is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1] Propynyl-PEG1-Ac is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-002-05794 Supplier Medchemexpress LLC Supplier No. HYW09614650MG
Propynyl-PEG1-Ac is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1] Propynyl-PEG1-Ac is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups