Learn More
Selleck Chemical LLC Propyphenazone (4-Isopropylantipyrine) S5309-25mg
SDP
Catalog No. 503109112
Encompass_Preferred
Propyphenazone(4-Isopropylantipyrine Isopropyrine) an analgesic and antipyretic agent is a selective cyclooxygenase-2 inhibitor with anti-inflammatory activity
Catalog No. 50-310-9112 Supplier Selleck Chemical LLC Supplier No. S530925MG
Propyphenazone(4-Isopropylantipyrine Isopropyrine) an analgesic and antipyretic agent is a selective cyclooxygenase-2 inhibitor with anti-inflammatory activity