Learn More
Medchemexpress LLC Propyphenazone-d3 | 162935-29-7 | 99.0% | C14H15D3N2O | 1 MG
SDP
Catalog No. 5000432471
Encompass_Preferred
Propyphenazone-d3 is the deuterium labeled Propyphenazone. Propyphenazone is a pyrazolone derivative with anti-inflammatory, analgesic and antipyretic activity. Propyphenazone-based analogues act as proagents and selective cyclooxygenase-2 inhibitors. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32471 Supplier Medchemexpress LLC Supplier No. HYA0273S1MG
Add to Cart
Add to Cart
Propyphenazone-d3 is the deuterium labeled Propyphenazone. Propyphenazone is a pyrazolone derivative with anti-inflammatory, analgesic and antipyretic activity. Propyphenazone-based analogues act as proagents and selective cyclooxygenase-2 inhibitors. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS