Learn More
Aviva Systems Biology Corp PROS1 Antibody - middle region (ARP56097_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56097_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PROS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PROS1. Alternate names: PROS, PS 26, PS21, PS22, PS23, PS24, PS25, PSAApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61716