Learn More
MSC PROSAFE PVC FLR MARK TAPE
Catalog No. 5000674633
Encompass
This Pro-Safe floor and aisle marking tape is made from polyvinylchloride and features a red and white striped pattern. It is 3 in. wide, 100 ft. long, and 30 mil thick, making it suitable for general traffic areas.
  • Red and white striped pattern
  • Non anti-slip surface
  • Polyvinylchloride material
  • Width of 3 in.
  • Length of 100 ft.
  • Thickness of 30 mil
  • Designed for general traffic
  • Keeps facility neat and organized
  • Prop 65 compliant
Catalog No. 50-006-74633 Supplier MSC Supplier No. 58281890
This Pro-Safe floor and aisle marking tape is made from polyvinylchloride and features a red and white striped pattern. It is 3 in. wide, 100 ft. long, and 30 mil thick, making it suitable for general traffic areas.
  • Red and white striped pattern
  • Non anti-slip surface
  • Polyvinylchloride material
  • Width of 3 in.
  • Length of 100 ft.
  • Thickness of 30 mil
  • Designed for general traffic
  • Keeps facility neat and organized
  • Prop 65 compliant