Learn More
Medchemexpress LLC Prosaikogenin D | 103629-72-7 | 10 MG
SDP
Catalog No. 5000417569
Encompass_Preferred
Prosaikogenin D is isolated from the roots of Buleurum bicaule Helm (Apiaceae) and exhibits anti-cancer activity.
  • Suppresses the proliferation of A549 cells with an IC50 of 57.2 μg/mL.
Catalog No. 50-004-17569 Supplier Medchemexpress LLC Supplier No. HYN766710MG
Prosaikogenin D is isolated from the roots of Buleurum bicaule Helm (Apiaceae) and exhibits anti-cancer activity.
  • Suppresses the proliferation of A549 cells with an IC50 of 57.2 μg/mL.