Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prosapogenin A | 19057-67-1 | 10 MG
SDP

Catalog No. 5000418723
Encompass_Preferred

Prosapogenin A, also known as Progenin III, is a natural product isolated from Veratrum species. This compound is recognized for its potent anticancer properties, primarily through its ability to induce apoptosis in various human cancer cells. Its mechanism of action involves the inhibition of the STAT3 signaling pathway and glycolysis, crucial processes often dysregulated in cancer progression.

  • Induces apoptosis in human cancer cells.
  • Inhibits the STAT3 signaling pathway and glycolysis.
  • Arrests the cell cycle at G2/M phase in HepG2 cells.
  • Demonstrates antitumor efficacy in xenograft mouse models.
  • Induces GSDME-dependent pyroptosis in anaplastic thyroid cancer cells.
  • Molecular formula: C39H62O12.
  • Molecular weight: 722.90.
  • Store at 4°C, protected from light.

Catalog No. 50-004-18723 Supplier Medchemexpress LLC Supplier No. HYN694010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prosapogenin A, also known as Progenin III, is a natural product isolated from Veratrum species. This compound is recognized for its potent anticancer properties, primarily through its ability to induce apoptosis in various human cancer cells. Its mechanism of action involves the inhibition of the STAT3 signaling pathway and glycolysis, crucial processes often dysregulated in cancer progression.

  • Induces apoptosis in human cancer cells.
  • Inhibits the STAT3 signaling pathway and glycolysis.
  • Arrests the cell cycle at G2/M phase in HepG2 cells.
  • Demonstrates antitumor efficacy in xenograft mouse models.
  • Induces GSDME-dependent pyroptosis in anaplastic thyroid cancer cells.
  • Molecular formula: C39H62O12.
  • Molecular weight: 722.90.
  • Store at 4°C, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.