Learn More
Pro Scientific Inc Bio-Gen PRO200 Homogenizer, 144 Watts, Variable Speed Control, 120 V
SDP
Catalog No. NC9771464
Encompass_Preferred
The Bio-Gen PRO200 Homogenizer is a powerful, versatile tool designed for efficient sample processing in laboratories. It offers a hand-held or post-mounted operation, making it suitable for various tube sizes and small beakers. With a high-torque motor rated at 144 Watts and a broad speed range of 5,000 - 35,000 rpm, users can achieve rapid homogenization in seconds. Built in the USA and backed by a 2-year warranty, it’s a reliable choice for any scientific application.
Catalog No. NC9771464 Supplier Pro Scientific Inc Supplier No. 0101200
The Bio-Gen PRO200 Homogenizer is a powerful, versatile tool designed for efficient sample processing in laboratories. It offers a hand-held or post-mounted operation, making it suitable for various tube sizes and small beakers. With a high-torque motor rated at 144 Watts and a broad speed range of 5,000 - 35,000 rpm, users can achieve rapid homogenization in seconds. Built in the USA and backed by a 2-year warranty, it’s a reliable choice for any scientific application.