Learn More
Electron Microscopy Sciences ProScope C-Mount Adapter
SDP
Catalog No. 5033647
Encompass
Use your own optics with our industry standard 25.4mm c-mount adapter. Attach the ProScope to an existing microscope, telescope or camera lenses to capture digital images and expand your digital imaging tools.
Catalog No. 50-336-47 Supplier Electron Microscopy Sciences Supplier No. 6835310
Use your own optics with our industry standard 25.4mm c-mount adapter. Attach the ProScope to an existing microscope, telescope or camera lenses to capture digital images and expand your digital imaging tools.