Learn More
Electron Microscopy Sciences ProScope HR CSI Science Level 2
SDP
Supplier:  Electron Microscopy Sciences 6835061
Encompass
A handheld digital microscope that captures stills, video and time lapse. Since 2001 the ProScope HR has been a mainstay in science education, law enforcement, and manufacturing quality control worldwide. 30-day no questions asked return policy. Two year warranty. Kit contains: ProScope HR, 0-10x Lens, 50x Lens, 200x Lens, Precision Stand, Carrying Case.
Catalog No. 50-336-53