Learn More
Electron Microscopy Sciences ProScope Micro Mobile iPhone 4/4s kit
SDP
Catalog No. 501902710
Encompass
The new ProScope Micro Mobile is adaptable to mobile devies. Using a custom coated glass optics that works with your onboard camera, you can obtain superb quality images up to 80X. The built-in LEDs adjust to reduce reflection and are powered by a sleek rechargeable battery for up to 5 hours continuous use.
Made with impact resistant Nylon and ABS for years of field, lab, classroom and exam room use. There are adapters for use with camera-equipped smart phones and tablets.
The Micro Mobile features 20X to 80X magnification via Edmund pro level glass optics for better image quality and color reproduction and adjustable LEDs to reduce surface reflection. USB charging cable to recharge to full capacity is complete in 2 hours. Includes a built-in desktop stand. Kit includes Proscope, 20-80x lens, iPhone 4/4S sleeve, adjustable nylon lens cone, stand, and USB charging cable.
Catalog No. 50-190-2710 Supplier Electron Microscopy Sciences Supplier No. 6835103
The new ProScope Micro Mobile is adaptable to mobile devies. Using a custom coated glass optics that works with your onboard camera, you can obtain superb quality images up to 80X. The built-in LEDs adjust to reduce reflection and are powered by a sleek rechargeable battery for up to 5 hours continuous use.
Made with impact resistant Nylon and ABS for years of field, lab, classroom and exam room use. There are adapters for use with camera-equipped smart phones and tablets.
The Micro Mobile features 20X to 80X magnification via Edmund pro level glass optics for better image quality and color reproduction and adjustable LEDs to reduce surface reflection. USB charging cable to recharge to full capacity is complete in 2 hours. Includes a built-in desktop stand. Kit includes Proscope, 20-80x lens, iPhone 4/4S sleeve, adjustable nylon lens cone, stand, and USB charging cable.