Learn More
Electron Microscopy Sciences ProScope Stand
SDP
Catalog No. 5033663
Encompass
Designed and developed for law enforcement, the ProScope Stand will steadily hold the ProScope in a wide variety of positions. Also for industrial inspection and biology.
Catalog No. 50-336-63 Supplier Electron Microscopy Sciences Supplier No. 6835313
Designed and developed for law enforcement, the ProScope Stand will steadily hold the ProScope in a wide variety of positions. Also for industrial inspection and biology.