Learn More
Electron Microscopy Sciences ProScope ProScope T1 WiFi Microscope without lens
SDP
Supplier:  Electron Microscopy Sciences 6835000T1
Encompass
Powered by a rechargable battery, ProScope T1 WiFi Microscope creates its own Wi-Fi network. It can transmit images simultaneously to as many as 253 devices (IOS or Android). With these apps, users can view live images, freeze and capture them to the photo album in their device.
Catalog No. 50-193-1657