Learn More
Cayman Chemical Prostaglandin F2α 1,11-lactone, Purity: ≥98%, CAS: 62410-84-8, 10 mg
SDP
Catalog No. 502972488
Encompass_Preferred
PGF2α 1,11-lactone is a lipid-soluble internal ester of PGF2α. Hydrolysis of the lactone by plasma esterases readily produces free PGF2α in rats and monkeys, but not in humans.{1170} PGF2α 1,11-lactone is active as an antifertility agent with greatly reduced vasoactivity compared to PGF2α.{1170}
Catalog No. 50-297-2488 Supplier Cayman Chemical Supplier No. 1611010 MG
PGF2α 1,11-lactone is a lipid-soluble internal ester of PGF2α. Hydrolysis of the lactone by plasma esterases readily produces free PGF2α in rats and monkeys, but not in humans.{1170} PGF2α 1,11-lactone is active as an antifertility agent with greatly reduced vasoactivity compared to PGF2α.{1170}