Learn More
Cayman Chemical PROSGlnDN F2a 1 15lacton 10mg
SDP
Catalog No. 502973186
Encompass_Preferred
PGF2α 1,15-lactone is a lipid-soluble internal ester of PGF2α. Hydrolysis of the lactone readily produces free PGF2α in plasma. In rhesus monkeys, a total dose of 15 mg of PGF2α 1,15-lactone terminates EArly pregnancy, whereas PGF2α is ineffective.{4371}
Catalog No. 50-297-3186 Supplier Cayman Chemical Supplier No. 1612010 MG
PGF2α 1,15-lactone is a lipid-soluble internal ester of PGF2α. Hydrolysis of the lactone readily produces free PGF2α in plasma. In rhesus monkeys, a total dose of 15 mg of PGF2α 1,15-lactone terminates EArly pregnancy, whereas PGF2α is ineffective.{4371}