Learn More
Cayman Chemical Prostaglandin F2α diethyl amide, Purity: ≥98%, 10 mg
SDP
Catalog No. 502968827
Encompass_Preferred
An analog of PGF2α in which the C-1 carboxyl group has been modified to an N-diethyl that is inert to coneal amidase activity
Catalog No. 50-296-8827 Supplier Cayman Chemical Supplier No. 1602310 MG
An analog of PGF2α in which the C-1 carboxyl group has been modified to an N-diethyl that is inert to coneal amidase activity