Learn More
Description
- Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.
- Made with a microporous film that is laminated to a spundbonded polypropylene substrate
- Front zipper closure
- Elastic wrists, ankles
- Attached hood
- Serged seams
- Collar
- White
- Thickness: 10 mil
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | 6X-Large |
| Gender | Unisex |
| Material | ProShield |
| Color | White |
| Disposable | Single-use |
| Waist Style | Non-elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.