Learn More
DuPont™ ProShield™ 6 SFR Coverall (all)
Catalog No. 17500155 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
5X-Large
6X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-155 5X-Large
17-500-156 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-500-155 Supplier DuPont™ Supplier No. TM120SBU5X002500
Add to Cart
Add to Cart
This item is not returnable. View return policy
SFR protective collared coveralls help extend the life of your flame resistant garments.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Laydown collar
  • Open wrists
  • Open ankles
  • Manufactured under specifications that do not contain natural rubber latex
  • Provides a barrier against non-hazardous particles and aerosols
TRUSTED_SUSTAINABILITY
Specifications
Size 5X-Large
Gender Unisex
Material Proshield
Color Blue
Disposable Single-use
Ankle Style Open
Cuff Style Open
Neck Style Collared
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Flame, Particle
No. of Pockets None