Learn More
MSC PRO-SOURCE Floor Polishing Pad, Fine, Polyester, 15 inch
Catalog No. 502839775
Encompass
PRO-SOURCE floor polishing pads are designed for general cleaning and polishing soft waxes on wood floors. They are fine-grade, made from polyester, and have a 15-inch diameter.
  • Fine grit grade
  • Polyester material
  • 15 inch diameter
  • Suitable for general cleaning
  • Ideal for polishing soft waxes on wood floors
  • Tan color
  • Sold in packs of five
Catalog No. 50-283-9775 Supplier MSC Supplier No. 78318441
PRO-SOURCE floor polishing pads are designed for general cleaning and polishing soft waxes on wood floors. They are fine-grade, made from polyester, and have a 15-inch diameter.
  • Fine grit grade
  • Polyester material
  • 15 inch diameter
  • Suitable for general cleaning
  • Ideal for polishing soft waxes on wood floors
  • Tan color
  • Sold in packs of five