Learn More
MSC ProSource Caster Wheels, 3 in, Plate Mount, Swivel, Steel
Catalog No. 5000529268
Encompass
These caster wheels feature rugged construction and a swivel style, designed with a 3 inch diameter, 1.7 inch width, and 4 inch overall height. They feature a plate mount and plain bearing type, with a hard wheel hardness.
  • Rugged construction
  • Swivel style
  • 3 in wheel diameter
  • Plate mount type
  • Plain bearing type
  • 1.7 in wheel width
  • Hard wheel hardness
  • 280 load capacity
  • 4 in overall height
  • Steel wheel material
  • Black wheel color
  • No brake/locking style
  • Back-mounted design
Catalog No. 50-005-29268 Supplier MSC Supplier No. 06505127
These caster wheels feature rugged construction and a swivel style, designed with a 3 inch diameter, 1.7 inch width, and 4 inch overall height. They feature a plate mount and plain bearing type, with a hard wheel hardness.
  • Rugged construction
  • Swivel style
  • 3 in wheel diameter
  • Plate mount type
  • Plain bearing type
  • 1.7 in wheel width
  • Hard wheel hardness
  • 280 load capacity
  • 4 in overall height
  • Steel wheel material
  • Black wheel color
  • No brake/locking style
  • Back-mounted design