Learn More
MSC Continental Huskee Round Receptacle, 20 gal, Gray, NSF, 1 ea
Catalog No. 502866469
Encompass
This Huskee round receptacle is a durable, commercial-grade container designed for various uses, from refuse collection to food preparation. Molded with seamless construction, it offers strength and longevity for demanding environments.
  • Molded with seamless construction for strength and longevity
  • Heavy-duty handles for secure lifting and moving
  • Reinforced base for enhanced durability and dolly compatibility
  • Made of high-quality LLDPE resin, safe for food storage
  • NSF Standard 2 certified, OSHA compliant, and USDA accepted equipment
  • Stackable design for efficient storage
Catalog No. 50-286-6469 Supplier MSC Supplier No. 66506247
This Huskee round receptacle is a durable, commercial-grade container designed for various uses, from refuse collection to food preparation. Molded with seamless construction, it offers strength and longevity for demanding environments.
  • Molded with seamless construction for strength and longevity
  • Heavy-duty handles for secure lifting and moving
  • Reinforced base for enhanced durability and dolly compatibility
  • Made of high-quality LLDPE resin, safe for food storage
  • NSF Standard 2 certified, OSHA compliant, and USDA accepted equipment
  • Stackable design for efficient storage