Learn More
MSC PRO-SOURCE Plastic Trigger Sprayer, White, 9-1/4 in, 1 ea
Catalog No. 502867943
Encompass
PRO-SOURCE plastic trigger sprayers are designed for dispensing liquids from bottles. They feature a leakproof screw closure and multiple spray settings for versatile use.
  • Made of plastic
  • For use with 32 oz bottles
  • White color
  • Has a 9-1/4 in dip tube length
  • Features mist, spray, and off settings
  • Leakproof screw closure
  • Provides 0.70 mL output
Catalog No. 50-286-7943 Supplier MSC Supplier No. 31018864
PRO-SOURCE plastic trigger sprayers are designed for dispensing liquids from bottles. They feature a leakproof screw closure and multiple spray settings for versatile use.
  • Made of plastic
  • For use with 32 oz bottles
  • White color
  • Has a 9-1/4 in dip tube length
  • Features mist, spray, and off settings
  • Leakproof screw closure
  • Provides 0.70 mL output