Learn More
MSC PRO-SOURCE Plastic Trigger Sprayer, 9-1/4 in Dip Tube, 1 ea
Catalog No. 502817025
Encompass
The PRO-SOURCE plastic trigger sprayer is designed for use with 32 oz bottles, offering mist, spray, and off settings. It features a leakproof screw closure for secure operation.
  • Made from durable plastic material
  • Equipped with a 9-1/4 in dip tube length
  • Provides an output of 0.80 mL per spray
  • Features a 28;400 screw closure size for compatibility
  • Includes mist, spray, and off settings for versatile use
  • Designed with a leakproof screw closure
Catalog No. 50-281-7025 Supplier MSC Supplier No. 31018856
The PRO-SOURCE plastic trigger sprayer is designed for use with 32 oz bottles, offering mist, spray, and off settings. It features a leakproof screw closure for secure operation.
  • Made from durable plastic material
  • Equipped with a 9-1/4 in dip tube length
  • Provides an output of 0.80 mL per spray
  • Features a 28;400 screw closure size for compatibility
  • Includes mist, spray, and off settings for versatile use
  • Designed with a leakproof screw closure