Learn More
Thermo Scientific™ ProSpecT™ C. difficile Toxin A/B Microplate Assay
GSA_VA
Catalog No. R244596 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
96 Wells/Kit
Catalog No. Quantity
R244596 96 Wells/Kit
1 options
This item is not returnable. View return policy
Catalog No. R244596 Supplier Thermo Scientific™ Supplier No. R244596
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rapidly detect Clostridium difficile toxin A and B from human fecal samples.
Rapidly detect Clostridium difficile toxin A and B in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ C. difficile Toxin A/B Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with the first C. difficile Toxin A/B assay to combine excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT C. difficile Toxin A/B is a direct, qualitative enzyme immmunoassay for the detection of Clostridium difficile toxin A and B and is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures, and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT C. difficile Toxin A/B provides same-day toxin detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description ProSpecT C. difficile Toxin A/B Microplate Assay
Includes Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity 96 Wells/Kit