Learn More
Thermo Scientific™ ProSpecT™ Cryptosporidium Microplate Assay
GSA_VA
Catalog No. R2454096 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
96 Wells/Kit
Catalog No. Quantity
R2454096 96 Wells/Kit
1 options
This item is not returnable. View return policy
Catalog No. R2454096 Supplier Thermo Scientific™ Supplier No. R2454096
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rapidly detect Crytosporidium specific antigen from human fecal samples.
Rapidly detect Crytosporidium specific antigens (CSA) in fresh or preserved fecal specimens using Thermo Scientific™ ProSpecT™ Cryptosporidium Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Crytosporidium. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT Cryptosporidium is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures, and common reagents, maximizing operational efficiency, productivity and throughput.
ProSpecT Cryptosporidium provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description ProSpecT Cryptosporidium Microplate Assay
Detectable Analytes Cryptosporidium Antigen
Includes Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity 96 Wells/Kit