Learn More
Description
Rapidly detect specific antigens in human specimens using Thermo Scientific™ ProSpecT™ Microplate Assays with the Thermo Scientific ProSpecT Microplate Assay Wash Buffer. Its concentration is 10X.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | ProSpecT Microplate Assay Wash Buffer |
| CE Marker | Yes |
| Concentration | 10X |
| Format | Bottle |
| Quantity | 1L/Bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.