Learn More
MESA LABS BOZEMAN MFG FACILITY ProSpore 4ml Ampoule 10/pk
SDP
Catalog No. NC9605165
Encompass
Hermetically sealed, type I borosilicate glass ampoule. The ampoule is filled with a modified Soybean Casein Digest Broth containing bromocresol purple acid indicator. Each ampoule also contains a population of G. stearothermophilus spores. ProSpore Ampoule (4 ml) has received 510(k) notification for sale to healthcare facilities with a 48-hour incubation period. Growth is evident by either turbidity and/or a color change from purple to or toward yellow. Expiration is 18 months from the date of manufacture.
This item is not returnable. View return policy
Catalog No. NC9605165 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. 7702PS410
Hermetically sealed, type I borosilicate glass ampoule. The ampoule is filled with a modified Soybean Casein Digest Broth containing bromocresol purple acid indicator. Each ampoule also contains a population of G. stearothermophilus spores. ProSpore Ampoule (4 ml) has received 510(k) notification for sale to healthcare facilities with a 48-hour incubation period. Growth is evident by either turbidity and/or a color change from purple to or toward yellow. Expiration is 18 months from the date of manufacture.