Learn More
MESA LABS BOZEMAN MFG FACILITY Mesa Labs ProSpore Ampoule, Self-Contained Biological Indicator, No Activation Required, 4 mL
SDP
Catalog No. NC3314477
Encompass
The Mesa Labs ProSpore Ampoule is a revolutionary self-contained biological indicator designed for validating steam sterilization processes. It features a unique 4 mL glass ampoule with a population of *Geobacillus stearothermophilus* spores, allowing for easy monitoring of sterilization effectiveness. With no activation required, results are obtained within 48 hours at reduced incubation temperature ranges of 55–60 °C. This product excels in reliability for both healthcare and industrial applications, making it a trusted choice for ensuring sterilization success.
Catalog No. NC3314477 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. 7702PS550
The Mesa Labs ProSpore Ampoule is a revolutionary self-contained biological indicator designed for validating steam sterilization processes. It features a unique 4 mL glass ampoule with a population of *Geobacillus stearothermophilus* spores, allowing for easy monitoring of sterilization effectiveness. With no activation required, results are obtained within 48 hours at reduced incubation temperature ranges of 55–60 °C. This product excels in reliability for both healthcare and industrial applications, making it a trusted choice for ensuring sterilization success.