Learn More
Enzo Life Sciences Urinary Prostacyclin ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900025
Encompass_Preferred
The Urinary Prostacyclin EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 2 by measuring the stable metabolites in one simple assay. Alternative Name: PGI2. Prostaglandin I2. Sensitivity: 6.58 pg/ml (range 7.81 - 2,000 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-109