Learn More
Enzo Life Sciences Prostaglandin A2 (10mg). CAS: 13345-50-1
SDP
Supplier:  Enzo Life Sciences BMLPG0020010
Encompass_Preferred
Blocks cell cycle at the G1 and G2/M phase [1] by inhibiting G1 phase cyclin dependent kinases [2] downregulating cyclin D1 [3]. Modulates all three aspects of the glutathione-mediated biotransformation system, i.e. GSH levels, glutathione S-transferase activity and transport of GSH conjugates [4]. Antitumor activity. Induces γ-glutamylcysteine synthetase and apoptosis in L-1210 cells. Alternative name: Medullin, Prosta-5,10,13-trien-1-oic acid, 15-hydroxy-9-oxo-, (5Z,13E,15S)-, PGA2. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol, acetone or ethyl acetate. Long Term Storage: -20°C.
Catalog No. 50-201-1411