Learn More
Cayman Chemical Prostaglandin D2, 41598-07-6, 1 mg
SDP
Supplier:  Cayman Chemical 120101
Encompass_Preferred
Molecular Formula C20H32O5, Purity ≥98%, Formula Weight 352.5, Synonyms: PGD2; The major eicosanoid product of mast cells released in large quantities during allergic and asthmatic anaphylaxis; also produced in the brain via an alternative pathway involving a soluble, secreted PGD-synthase also known as -trace where it induces normal physiological sleep and lowering of body temperature; inhibits platelet aggregation, relaxes vascular smooth muscle, and inhibits human ovarian tumor cell proliferation (IC50 = 6.8 M).
Catalog No. NC9783924