Learn More
Cayman Chemical Prostaglandin E Metabolite ELISA Kit, 96 solid wells
SDP
Supplier:  Cayman Chemical 51453196SOLIDWELLS
Encompass_Preferred
Because of the rapid metabolism of PGE2, the determination of in vivo PGE2 biosynthesis is often best accomplished by the measurement of PGE2 metabolites. Cayman’s PGEM Assay converts all 13,14-dihydro-15-keto PGE2 and 13,14-dihydro-15-keto PGA2 into a single stable derivative, which is easily measurable by ELISA. This assay is therefore the method of choice if the samples in question have undergone extensive metabolism prior to collection.
Catalog No. NC0960078