Learn More
STEMCELL Technologies Prostaglandin E2, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 72194
Encompass
Prostaglandin E₂ (PGE₂) is the most biologically active and well-studied prostaglandin. It binds with very high affinity to the EP1, EP2, EP3, and EP4 receptors (Ki = 9.1, 4.9, 0.33, 0.79 nM respectively). (Abramovitz et al., Bos et al.)MAINTENANCE AND SELF-RENEWAL· Required for the development of hematopoietic stem cells (HSCs) in mice and zebrafish (North et al.).· Improves engraftment of mouse HSCs, possibly through increasing homing, survival, and/or self-renewal (Hoggatt et al. 2009, Hoggatt et al. 2013, North et al.).DIFFERENTIATION· Promotes differentiation of hematopoietic progenitor cells from mouse, macaque, and human embryonic stem cells (Gori et al., North et al., Woods et al.).· Promotes differentiation of myeloid-derived suppressor cells from hematopoietic progenitors (Sinha et al.).· Promotes differentiation of Th17 cells from naïve T-cells (Boniface et al.).
Catalog No. NC0661663