Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Prostaglandin E2 Competitive ELISA Kit

Catalog No. ENEHPGE2
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENEHPGE2 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENEHPGE2 Supplier Invitrogen™ Supplier No. EHPGE2
Only null left
Add to Cart
Add to Cart

ELISA

The Human Prostaglandin E2 (Hu PGE2) ELISA quantitates Hu PGE2 in human serum, whole blood, saliva, urine or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu PGE2. Principle of the method The Human PGE2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is inversely proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Prostaglandin E2 (PGE2) is a lipid mediator derived from arachidonic acid through the actions of cyclooxygenase (COX) enzymes and terminal prostaglandin E synthases (PGES). It plays a significant role in mediating inflammation, pain sensation, and fever by acting on four receptor subtypes: EP1, EP2, EP3, and EP4. PGE2's involvement in inflammatory responses is well-documented, particularly in diseases such as rheumatoid arthritis and osteoarthritis. Various medications, including nonsteroidal anti-inflammatory drugs (NSAIDs) and selective COX-2 inhibitors, are used to reduce PGE2 production, thereby alleviating inflammation and pain. However, these drugs can have side effects, such as gastrointestinal issues and increased risk of thrombosis. Additionally, PGE2 has a critical role in immune regulation, affecting T cell function and, by extension, the adaptive immune response. It also has a significant impact on skeletal disorders and pain through mechanisms that are still being explored.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 39.1 to 2500 pg/mL
Assay Sensitivity 13.4 pg/mL
Description PGE2 Competitive ELISA Kit
Conjugate AP
Immunoassay Kit Format Colorimetric ELISA (p-NPP substrate) with Plates
Product Type ELISA
Sample Type Saliva, Serum, Supernatant, Urine, Whole Blood
Specificity No significant cross-reaction with 13,14-dihydro-15-keto-PGF2a; 6,15-keto-13-dihydro-PGF1a; Thromboxane B2; 2-Arachidonoylglycerol; Anandamide; PGD2; Arachadonic acid. Less than 1% cross-reaction with PFG2a; 6-keto-PGF1a; PGA2; PGB1; Approx. 16% cross-rea
Sufficient For 96 microplate wells
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 4%
Intraassay CV 10.7
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent, Detection Antibody, AP Conjugate, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye AP
Measurement Absorbance at 405 nm minus absorbance (at 570 nm)
Precision Dependent upon sample type and level
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Saliva 10 μL, Serum 10 μL, Supernatant 100 μL, Urine 10 μL, Whole Blood 10 μL
Storage Requirements -20°C
Target PGE2
Test Time 1 hrs 20 mins
Total Assay Time 2 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.