Learn More
Selleck Chemical LLC Prostaglandin E2 (PGE2) 5mg 363-24-6 Dinoprostone
SDP
Supplier:  Selleck Chemical LLC S30035MG
Encompass_Preferred
Prostaglandin E2 (PGE2, Dinoprostone) palys important effects in labour (softens cervix and causes uterine contraction) and also stimulates osteoblasts to release factors that stimulate bone resorption by osteoclasts. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8082