Learn More
Cayman Chemical Prostaglandin F2α, 551-11-1, 5 mg
SDP
Supplier:  Cayman Chemical 160105
Encompass_Preferred
A widely distributed primary PG occuring in many species; causes contraction of vascular, bronchial, intestinal, and myometrial smooth muscle, and also exhibits potent luteolytic activity
Catalog No. NC1134989