Learn More
Enzo Life Sciences Urinary Prostacyclin ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901025
Encompass_Preferred
The Urinary Prostacyclin EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. Quantitate PGI2 by measuring the stable metabolites in one simple assay. Alternative name: PGI2, Prostaglandin I2. Sensitivity: 6.58 pg/ml (range 7.81 - 2,000 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent . Content: DxS Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Catalog No. 50-200-6888