Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Prostasin/PRSS8 ELISA Kit

Catalog No. p-7230775
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHPRSS8 96 Tests
EHPRSS8X10 10 x 96 Tests
EHPRSS8X5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHPRSS8 Supplier Invitrogen™ Supplier No. EHPRSS8
Only null left
Add to Cart
Add to Cart

ELISA

The Human Prostasin/PRSS8 ELISA quantitates Hu Prostasin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Prostasin. Principle of the method The Human Prostasin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is highly expressed in prostate epithelia and is one of several proteolytic enzymes found in seminal fluid. The proprotein is cleaved to produce a light chain and a heavy chain which are associated by a disulfide bond. It is active on peptide linkages involving the carboxyl group of lysine or arginine.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16651
Assay Range 32.77 to 8000 pg/mL
Assay Sensitivity 30 pg/mL
Calibration Range 30 pg/mL to 8000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Cell Structure & Organelle Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human ANGPTL3, beta IG-H3, CA9, Cathepsin B, CD23, CHI3L1, CTLA-4, DKK-4, DPPIV, EDA-A2, Erythropoietin R, FGF-6, FGF-9, Gas 1, GITR, IGFBP-5, IL-1 F5, IL-1 F6, IL-1 F7, IL-1 F8, IL-1 F9, IL-1 F10, IL-1 R5, IL-21, IL-18, IL-20, IL-34, IL-5 R alpha, IL-10 R alpha, Layilin, Leptin R, Marapsin, Mer, MMP-7, P-Cadherin, PSMA, SIGIRR, TGF beta R3, Coagulation Factor III, TWEAK.
Sufficient For 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5652
Gene Symbol PRSS8
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Cell Cycle & Proliferation
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym CAP1, PROSTASIN
Target Prostasin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.