Learn More
Apexbio Technology LLC prostate apoptosis response protein PAR-4 (2-7) [Homo sapiens] 1mg
SDP
Catalog No. 5000096894
Encompass_Preferred
Prostate apoptosis response protein PAR-4 2-7 is a short peptide targeting transcriptional regulation pathways It is designed to modulate transcription factor activity thereby influencing apoptosis mechanisms particularly in prostate cells PAR-4 2-7 exerts its biological activity primarily through interaction with transcription factors via a leucine zipper zinc finger domain facilitating apoptosis in transformed or malignant cells while sparing normal cells Based on these properties PAR-4 2-7 holds research potential in studies investigating tumor suppression mechanisms and transcription-mediated apoptosis regulation in oncology and therapeutic research
Catalog No. 50-000-96894 Supplier Apexbio Technology LLC Supplier No. A10851
Prostate apoptosis response protein PAR-4 2-7 is a short peptide targeting transcriptional regulation pathways It is designed to modulate transcription factor activity thereby influencing apoptosis mechanisms particularly in prostate cells PAR-4 2-7 exerts its biological activity primarily through interaction with transcription factors via a leucine zipper zinc finger domain facilitating apoptosis in transformed or malignant cells while sparing normal cells Based on these properties PAR-4 2-7 holds research potential in studies investigating tumor suppression mechanisms and transcription-mediated apoptosis regulation in oncology and therapeutic research